(4-Bromophenyl)acetone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Bromophenylacetone

A19028-06 A19028-14
CAAAA19028-06EA 140.46 CAD
CAAAA19028-06 CAAAA19028-14
(4-Bromophenyl)acetone 98%
(4-Bromophenyl)acetone

Purity: 98% CAS No.: 6186-22-7 Molecular Formula: C9H9BrO Formula Weight: 213.07 MDL No.: MFCD00210401

Formula: C₉H₉BrO
MW: 213.07
Storage Temperature: Ambient
MDL Number: MFCD00210401
CAS Number: 6186-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR