Thionin(acetate)

Supplier: Thermo Scientific Chemicals
Warning

A18912-06 A18912-14
CAAAA18912-06EA 107.16 CAD
CAAAA18912-06 CAAAA18912-14
Thionin(acetate)
Thionin (Acetate)

Used as a biogical stain.


CAS No.: 78338-22-4 Molecular Formula: C14H13N3O2S Formula Weight: 287.34 MDL No.: MFCD00081194


Used as a biogical stain

Formula: C₁₄H₁₃N₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD00081194
CAS Number: 78338-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR