2,5-Piperazinedione 99%
Supplier: Thermo Scientific Chemicals
A18822-14
A18822-22
CAAAA18822-14EA
91.64
CAD
CAAAA18822-14
CAAAA18822-22
2,5-Piperazinedione 99%
2,5-Piperazinedione
Purity: 99%
CAS No.: 106-57-0
Molecular Formula: C4H6N2O2
Formula Weight: 114.10
Melting Point: ca 300° dec.
Beilstein Reference: 112112
Merck Reference: 14,7466
MDL No.: MFCD00006009
Formula:
C₄H₆N₂O₂ Melting Pt: ∼ 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006009 CAS Number: 106-57-0 EINECS: 203-411-5 Merck Index: 14,7466 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...