4'-Hydroxy-3'-methylacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18540-03
A18540-06
A18540-14
CAAAA18540-03EA
48.19
CAD
CAAAA18540-03
CAAAA18540-06
CAAAA18540-14
4'-Hydroxy-3'-methylacetophenone 98%
4'-Hydroxy-3'-methylacetophenone
Purity: 98%
CAS No.: 876-02-8
Molecular Formula: C9H10O2
Formula Weight: 150.18
Melting Point: 107-109°
Boiling Point: 175°/1mm
MDL No.: MFCD00002231
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 175 °C (1 mmHg) Melting Pt: 107…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002231 CAS Number: 876-02-8 EINECS: 212-880-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...