Sodium 2,4-dinitrobenzenesulfonate 97%

Supplier: Thermo Scientific Chemicals

A18493-06 A18493-14
CAAAA18493-06EA 45.66 CAD
CAAAA18493-06 CAAAA18493-14
Sodium 2,4-dinitrobenzenesulfonate 97%
Sodium 2,4-dinitrobenzenesulphonate

Purity: 97% CAS No.: 885-62-1 Molecular Formula: C6H3N2NaO7S Formula Weight: 270.15 Beilstein Reference: 6676807 MDL No.: MFCD00007471

Formula: C₆H₃N₂NaO₇S
Storage Temperature: Ambient
MDL Number: MFCD00007471
CAS Number: 885-62-1
EINECS: 212-943-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR