4,4'-Bis(dimethylamino)diphenylmethane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18466-18
A18466-30
CAAAA18466-18EA
81.05
CAD
CAAAA18466-18
CAAAA18466-30
4,4'-Bis(dimethylamino)diphenylmethane 98+%
4,4'-Bis(dimethylamino)diphenylmethane
Purity: 98+%
CAS No.: 101-61-1
Molecular Formula: C17H22N2
Formula Weight: 254.38
Flash Point: 178°(352°F)
Melting Point: 85-88°
Boiling Point: 155-157°/0.1mm
Beilstein Reference: 479956
Merck Reference: 14,6176
UN No.: 3077
MDL No.: MFCD00008317
Formula:
C₁₇H₂₂N₂ Boiling Pt: 155…157 °C/0.1 mm Melting Pt: 85…88 °C Flash Pt: 178 °C(352 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008317 CAS Number: 101-61-1 EINECS: 202-959-2 UN: 3077 ADR: "9",III Merck Index: 14,6176 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...