4'-Benzyloxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Benzyloxyacetophenone
A18302-06
A18302-14
CAAAAA18302-22EA
1229.6
CAD
CAAAAA18302-22
CAAAA18302-06
CAAAA18302-14
4'-Benzyloxyacetophenone 98%
4'-Benzyloxyacetophenone
Purity: 98%
CAS No.: 54696-05-8
Molecular Formula: C15H14O2
Formula Weight: 226.28
Melting Point: 91-92°
Beilstein Reference: 1912124
Formula:
C₁₅H₁₄O₂ MW: 226.27 Melting Pt: 91…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017247 CAS Number: 54696-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...