m-Cresol purple sodium salt

Supplier: Thermo Scientific Chemicals

Synonyms: m-Cresolsulfonphthalein sodium salt, m-Cresolsulphonphthalein sodium salt, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[d][1,2]oxathiole 2,2-dioxide, sodium salt

A18025-06 A18025-14
CAAAAA18025-06EA 101.59 CAD
CAAAAA18025-06 CAAAAA18025-14
m-Cresol purple sodium salt
m-Cresol purple sodium salt

CAS No.: 62625-31-4 Molecular Formula: C21H17NaO5S Formula Weight: 404.42 MDL No.: MFCD00010177

Formula: C₂₁H₁₇NaO₅S
MW: 404.42 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010177
CAS Number: 62625-31-4
EINECS: 263-656-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR