Butylphosphonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A17983-06 A17983-14
CAAAAA17983-06EA 232.26 CAD
CAAAAA17983-06 CAAAAA17983-14
Butylphosphonic acid 98%
Butylphosphonic acid

Purity: 98% CAS No.: 3321-64-0 Molecular Formula: C4H11O3P Formula Weight: 138.10 Odor: Odorless Melting Point: 105-107° Beilstein Reference: 1749026 UN No.: 3261 MDL No.: MFCD00012295

Formula: C₄H₁₁O₃P
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00012295
CAS Number: 3321-64-0
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR