Pyridoxal hydrochloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl
A17855-06
A17855-14
A17855-22
CAAAA17855-06EA
108.7
CAD
CAAAA17855-06
CAAAA17855-14
CAAAA17855-22
Pyridoxal hydrochloride 99%
Pyridoxal hydrochloride
Purity: 99%
CAS No.: 65-22-5
Molecular Formula: C8H9NO3.HCl
Formula Weight: 203.63
Storage Temperature: 2°C to 8°C
Odor: Odorless
Beilstein Reference: 3656994
Merck Reference: 14,7978
MDL No.: MFCD00012809
Formula:
C₈H₁₀ClNO₃ MW: 203.62 g/mol Melting Pt: 172…174 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00012809 CAS Number: 65-22-5 EINECS: 200-602-5 Merck Index: 14,7978 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...