Imidazole-4,5-dicarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17783-22
A17783-06
A17783-14
CAAAAA17783-22EA
669.92
CAD
CAAAAA17783-22
CAAAA17783-06
CAAAA17783-14
Imidazole-4,5-dicarboxylic acid 97%
Imidazole-4,5-dicarboxylic acid
Purity: 97%
CAS No.: 570-22-9
Molecular Formula: C5H4N2O4
Formula Weight: 156.10
Melting Point: ca 283° dec.
Beilstein Reference: 147774
Formula:
C₅H₄N₂O₄ Melting Pt: ca 283 °C (dec.) Density: 1.749 Storage Temperature: Ambient |
MDL Number:
MFCD00005200 CAS Number: 570-22-9 EINECS: 209-327-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...