5-Methyl-2-thenoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17737-06
A17737-14
CAAAA17737-06EA
79.95
CAD
CAAAA17737-06
CAAAA17737-14
5-Methyl-2-thenoic acid 98+%
5-Methyl-2-thenoic acid
Purity: 98+%
CAS No.: 1918-79-2
Molecular Formula: C6H6O2S
Formula Weight: 142.18
Melting Point: 136-138°
Beilstein Reference: 113857
Formula:
C₆H₆O₂S Melting Pt: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005439 CAS Number: 1918-79-2 EINECS: 217-640-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...