3-Cyclopentylpropionyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A17715-14 A17715-22
CAAAA17715-14EA 105.7 CAD
CAAAA17715-14 CAAAA17715-22
3-Cyclopentylpropionyl chloride 98%
3-Cyclopentylpropionyl chloride

Purity: 98% CAS No.: 104-97-2 Molecular Formula: C8H13ClO Formula Weight: 160.64 Flash Point: 84°(184°F) Refractive Index: 1.4645 Odor: Acrid Boiling Point: 199-200° Beilstein Reference: 1856952 UN No.: 3265 MDL No.: MFCD00001391

Formula: C₈H₁₃ClO
Boiling Pt: 199…200 °C
Density: 1.049
Flash Pt: 84 °C (184 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001391
CAS Number: 104-97-2
EINECS: 203-257-9
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR