2,4-Diaminophenol dihydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17697-14
A17697-22
CAAAA17697-14EA
82.63
CAD
CAAAA17697-14
CAAAA17697-22
2,4-Diaminophenol dihydrochloride 98+%
2,4-Diaminophenol dihydrochloride
Purity: 98+%
CAS No.: 137-09-7
Molecular Formula: C6H8N2O.2HCl
Formula Weight: 197.07
Melting Point: ca 280° dec.
Beilstein Reference: 3913728
Merck Reference: 14,2983
UN No.: 2811
MDL No.: MFCD00012979
Formula:
C₆H₁₀Cl₂N₂O Melting Pt: ca 280 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012979 CAS Number: 137-09-7 EINECS: 205-279-4 UN: 2811 ADR: "6.1",III Merck Index: 14,2983 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...