Adenine hydrochloride, (max. 5% H₂O) 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7H-Purin-6-amine hydrochloride, Purin-6-amine hydrochloride, 6-Aminopurine hydrochloride

A17622-06 A17622-14 A17622-22
CAAAA17622-06EA 43.13 CAD
CAAAA17622-06 CAAAA17622-14 CAAAA17622-22
Adenine hydrochloride, (max. 5% H₂O) 98+%
Adenine hydrochloride

Purity: 98+% CAS No.: 2922-28-3 Molecular Formula: C5H5N5.HCl Formula Weight: 171.59 Melting Point: ca 290° dec. Beilstein Reference: 4009585

Formula: C₅H₅N₅·HCl
MW: 171.59 g/mol
Melting Pt: ∼ 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150864
CAS Number: 2922-28-3
EINECS: 220-867-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR