Methylcyclopentadiene dimer tech. 90% stabilised, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: 200 ppm Stabilised with 4-tert-butylpyrocatechol (TBC)

A17596-AP A17596-0J A17596-AU
CAAAAA17596-APEA 88.48 CAD
CAAAAA17596-AP CAAAA17596-0J CAAAA17596-AU
Methylcyclopentadiene dimer tech. 90% stabilised, Technical Grade
Methylcyclopentadiene dimer

Purity: 90% CAS No.: 26472-00-4 Molecular Formula: C12H16 Formula Weight: 160.26 Flash Point: 26°(79°F) Refractive Index: 1.4980 Melting Point: ≈-50° Boiling Point: 198-201° UN No.: 3295 MDL No.: MFCD00082297

Formula: C₁₂H₁₆
MW: 160.26 g/mol
Boiling Pt: 198…201 °C
Melting Pt: < –50 °C
Density: 0.940
Flash Pt: 26 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082297
CAS Number: 26472-00-4
EINECS: 247-724-5
UN: 3295
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR