Metanil yellow
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17527-18
A17527-30
CAAAA17527-18EA
45.98
CAD
CAAAA17527-18
CAAAA17527-30
Metanil yellow
Metanil Yellow
Common Applications: as indicator. Transition interval: pH: 1.2 (red) to 2.3 (yellow)
CAS No.: 587-98-4
Molecular Formula: C18H14N3NaO3S
Formula Weight: 375.38
Odor: Odorless
pH: ~10
Beilstein Reference: 3831568
Merck Reference: 14,5928
MDL No.: MFCD00007486
Formula:
C₁₈H₁₄N₃NaO₃S Storage Temperature: Ambient |
MDL Number:
MFCD00007486 CAS Number: 587-98-4 EINECS: 209-608-2 Merck Index: 14,5928 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...