Chromotrope 2R

Supplier: Thermo Scientific Chemicals
Warning

A17519-14 A17519-22
CAAAA17519-14EA 70.63 CAD
CAAAA17519-14 CAAAA17519-22
Chromotrope 2R
Chromotrope 2R

CAS No.: 4197-07-3 Molecular Formula: C16H10N2Na2O8S2 Formula Weight: 468.37 MDL No.: MFCD00003955

Formula: C₁₆H₁₀N₂Na₂O₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00003955
CAS Number: 4197-07-3
EINECS: 224-085-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR