Azure B (azure I)

Supplier: Thermo Scientific Chemicals

Synonyms: C.I. 52010, N,N,N′-Trimethylthionin, Azure B, Trimethylthionine chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride, Methylene Azur B

A17508-06 A17508-14 A17508-22
CAAAA17508-06EA 67.62 CAD
CAAAA17508-06 CAAAA17508-14 CAAAA17508-22
Azure B (azure I)
Azure B (Azure I)

For differentiating cellular RNA and DNA in plant tissues.


CAS No.: 531-55-5 Molecular Formula: C15H16ClN3S Formula Weight: 305.83 Odor: Odorless Melting Point: ca 205° dec. Beilstein Reference: 3922630 Merck Reference: 14,6058 MDL No.: MFCD00011935

Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011935
CAS Number: 531-55-5
EINECS: 208-511-2
Merck Index: 14,6058

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR