Procaine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Diethylaminoethyl 4-aminobenzoate hydrochloride

A17485-18 A17485-30
CAAAA17485-18EA 51.51 CAD
CAAAA17485-18 CAAAA17485-30
Procaine hydrochloride 99%
Procaine hydrochloride

A sodium channel blocker


Purity: 99% CAS No.: 51-05-8 Molecular Formula: C13H20N2O2.HCl Formula Weight: 272.78 Odor: Odorless Melting Point: 155-157° Boiling Point: 195-196°/17mm Merck Reference: 14,7757 UN No.: 2811 MDL No.: MFCD00013000

Formula: C₁₃H₂₁ClN₂O₂
MW: 272.77 g/mol
Boiling Pt: 195…196 °C (17 mmHg)
Melting Pt: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00013000
CAS Number: 51-05-8
EINECS: 200-077-2
UN: 2811
ADR: 6.1 III
Merck Index: 14,7757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR