Isonicotinaldehyde oxime 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17371-14
A17371-22
CAAAA17371-14EA
77.89
CAD
CAAAA17371-14
CAAAA17371-22
Isonicotinaldehyde oxime 97%
Isonicotinaldehyde oxime
Purity: 97%
CAS No.: 696-54-8
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Melting Point: 130-133°
Beilstein Reference: 2079
Formula:
C₆H₆N₂O Melting Pt: 130…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006446 CAS Number: 696-54-8 EINECS: 211-796-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...