Diethyl propylphosphonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyl 1-propylphosphonate
A17105-09
A17105-18
CAAAA17105-09EA
87.22
CAD
CAAAA17105-09
CAAAA17105-18
Diethyl propylphosphonate 97%
Diethyl propylphosphonate
Purity: 97%
CAS No.: 18812-51-6
Molecular Formula: C7H17O3P
Formula Weight: 180.19
Refractive Index: 1.4190
Boiling Point: 94-96°/12mm
MDL No.: MFCD00015139
Formula:
C₇H₁₇O₃P Boiling Pt: 94…96 °C/12 mm Storage Temperature: Ambient |
MDL Number:
MFCD00015139 CAS Number: 18812-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...