5-Hydroxy-1-tetralone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one
A17034-06
A17034-14
CAAAA17034-03EA
180.48
CAD
CAAAA17034-03
CAAAA17034-06
CAAAA17034-14
5-Hydroxy-1-tetralone 99%
5-Hydroxy-1-tetralone
Purity: 99%
CAS No.: 28315-93-7
Molecular Formula: C10H10O2
Formula Weight: 162.19
Melting Point: 209-211°
Beilstein Reference: 2437410
Formula:
C₁₀H₁₀O₂ MW: 162.19 g/mol Melting Pt: 209…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001693 CAS Number: 28315-93-7 EINECS: 248-958-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...