5,5-Diphenylhydantoin 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17013-14
A17013-22
A17013-36
CAAAAA17013-14EA
38.08
CAD
CAAAAA17013-14
CAAAAA17013-22
CAAAAA17013-36
5,5-Diphenylhydantoin 99%
5,5-Diphenylhydantoin
Purity: 99%
CAS No.: 57-41-0
Molecular Formula: C15H12N2O2
Formula Weight: 252.27
Melting Point: 293-298°
Beilstein Reference: 384532
Merck Reference: 14,7322
MDL No.: MFCD00005264
Formula:
C₁₅H₁₂N₂O₂ Melting Pt: 293…298 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005264 CAS Number: 57-41-0 EINECS: 200-328-6 Merck Index: 14,7322 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...