1,2-Cyclohexanedione dioxime 97%
Supplier: Thermo Scientific Chemicals
A16788-06
A16788-14
A16788-22
CAAAA16788-06EA
96.06
CAD
CAAAA16788-06
CAAAA16788-14
CAAAA16788-22
1,2-Cyclohexanedione dioxime 97%
1,2-Cyclohexanedione dioxime
Purity: 97%
CAS No.: 492-99-9
Molecular Formula: C6H10N2O2
Formula Weight: 142.16
Melting Point: 181-188° dec.
Beilstein Reference: 1909047
Merck Reference: 14,6560
MDL No.: MFCD00001663
Formula:
C₆H₁₀N₂O₂ Melting Pt: 181…188 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00001663 CAS Number: 492-99-9 EINECS: 207-769-3 Merck Index: 14,6560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...