5-Chloro-2-mercaptobenzothiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

A16747-06 A16747-14
CAAAA16747-06EA 173.8 CAD
CAAAA16747-06 CAAAA16747-14
5-Chloro-2-mercaptobenzothiazole 98%
5-Chloro-2-mercaptobenzothiazole

Purity: 98% CAS No.: 5331-91-9 Molecular Formula: C7H4ClNS2 Formula Weight: 201.70 Melting Point: 198-200° MDL No.: MFCD00005783

Formula: C₇H₄ClNS₂
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00005783
CAS Number: 5331-91-9
EINECS: 226-235-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR