Cyclooctanone oxime 98+%

Supplier: Thermo Scientific Chemicals

A16672-09 A16672-18
CAAAA16672-09EA 107.6 CAD
CAAAA16672-09 CAAAA16672-18
Cyclooctanone oxime 98+%
Cyclooctanone oxime

Purity: 98+% CAS No.: 1074-51-7 Molecular Formula: C8H15NO Formula Weight: 141.22 Melting Point: 36-37° Boiling Point: 128-129°/14mm MDL No.: MFCD00191967

Formula: C₈H₁₅NO
MW: 141.21
Boiling Pt: 128…129 °C (14 mmHg)
Melting Pt: 36…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00191967
CAS Number: 1074-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR