(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Carbethoxymethyl)triphenylphosphonium bromide, (2-Ethoxy-2-oxoethyl)triphenylphosphonium bromide

A16347-14 A16347-22
CAAAA16347-14EA 52.93 CAD
CAAAA16347-14 CAAAA16347-22 CAAAA16347-36
(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98+%
(Ethoxycarbonylmethyl)triphenylphosphonium bromide

Purity: 98+% CAS No.: 1530-45-6 Molecular Formula: CH3CH2O2CCH2P(C6H5)3Br Formula Weight: 429.30 Melting Point: ca 150° dec. Beilstein Reference: 3581273

Formula: C₂₂H₂₂BrO₂P
MW: 429.30 g/mol
Melting Pt: ∼ 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011835
CAS Number: 1530-45-6
EINECS: 216-230-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR