1-(Trimethylsiloxy)cyclohexene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Cyclohex-1-en-1-yloxy)trimethylsilane

A16099-09 A16099-18
CAAAA16099-09EA 102.23 CAD
CAAAA16099-09 CAAAA16099-18 CAAAA16099-30
1-(Trimethylsiloxy)cyclohexene 98%
1-(Trimethylsiloxy)cyclohexene

Purity: 98% CAS No.: 6651-36-1 Molecular Formula: C9H18OSi Formula Weight: 170.33 Flash Point: 42°(107°F) Refractive Index: 1.4470 Boiling Point: 165° Beilstein Reference: 1859394 UN No.: 1993 MDL No.: MFCD00001541

Formula: C₉H₁₈OSi
MW: 170.33
Boiling Pt: 165 °C
Density: 0.883
Flash Pt: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001541
CAS Number: 6651-36-1
EINECS: 229-675-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR