1,4-Phenylenediacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16036-03
A16036-06
CAAAA16036-03EA
45.19
CAD
CAAAA16036-03
CAAAA16036-06
CAAAA16036-14
1,4-Phenylenediacetic acid 97%
1,4-Phenylenediacetic acid
Purity: 97%
CAS No.: 7325-46-4
Molecular Formula: C10H10O4
Formula Weight: 194.19
Odor: Odorless
Melting Point: 252-256°
Beilstein Reference: 1960342
MDL No.: MFCD00004354
Formula:
C₁₀H₁₀O₄ Melting Pt: 252…256 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004354 CAS Number: 7325-46-4 EINECS: 230-801-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...