6-Methoxy-1-tetralone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Methoxy-1,2,3,4-tetrahydro-1-naphthalenone, 6-Methoxy-3,4-dihydronaphthalen-1(2H)-one

A15967-06 A15967-14 A15967-22
CAAAA15967-06EA 36.81 CAD
CAAAA15967-06 CAAAA15967-14 CAAAA15967-22
6-Methoxy-1-tetralone 99%
6-Methoxy-1-tetralone

Purity: 99% CAS No.: 1078-19-9 Molecular Formula: C11H12O2 Formula Weight: 176.22 Melting Point: 77-80° Boiling Point: 171°/11mm Merck Reference: 14,6000 MDL No.: MFCD00001695

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 171 °C (11 mmHg)
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00001695
CAS Number: 1078-19-9
EINECS: 214-078-0
Merck Index: 14,6000

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR