Pyridinium p-toluenesulfonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridin-1-ium 4-methylbenzenesulfonate, Pyridin-1-ium 4-methylbenzenesulphonate

A15708-14 A15708-22 A15708-36
CAAAA15708-14EA 106.18 CAD
CAAAA15708-14 CAAAA15708-22 CAAAA15708-36
Pyridinium p-toluenesulfonate 98+%
Pyridinium p-toluenesulphonate

Fieser: 8,427 9,400 12,420 15,276 16,287 18,303


Purity: 98+% CAS No.: 24057-28-1 Molecular Formula: C12H13NO3S Formula Weight: 251.31 Melting Point: 116-120° Beilstein Reference: 3764305

Formula: C₁₂H₁₃NO₃S
MW: 251.31 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00013108
CAS Number: 24057-28-1
EINECS: 246-002-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR