Pyridinium tribromide tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

A15684-14 A15684-22 A15684-36
CAAAAA15684-14EA 72.84 CAD
CAAAAA15684-14 CAAAAA15684-22 CAAAAA15684-36
Pyridinium tribromide tech. 90%, Technical Grade
Pyridinium tribromide

Fieser: 1,967 5,568 6,499 9,399 10,333 15,276


Purity: 90% CAS No.: 39416-48-3 Molecular Formula: C5H5N.HBr3 Formula Weight: 319.82 Melting Point: 128-136° Beilstein Reference: 3690144 Merck Reference: 14,7973 UN No.: 3261 MDL No.: MFCD00013223

Formula: C₅H₆Br₃N
Melting Pt: 128…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013223
CAS Number: 39416-48-3
EINECS: 254-446-8
UN: 3261
ADR: "8",III
Merck Index: 14,7973

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR