4-Chloropyridine hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15628-06
A15628-14
A15628-22
CAAAAA15628-06EA
50.24
CAD
CAAAAA15628-06
CAAAAA15628-14
CAAAAA15628-22
4-Chloropyridine hydrochloride 98+%
4-Chloropyridine hydrochloride
Purity: 98+%
CAS No.: 7379-35-3
Molecular Formula: C5H4ClN.HCl
Formula Weight: 150.01
Flash Point: 198°(388°F)
Odor: Weak
pH: 2
Melting Point: ca 210° subl.
Beilstein Reference: 3684904
MDL No.: MFCD00012829
Formula:
C₅H₅Cl₂N Melting Pt: ca 210 °C (subl.) Flash Pt: 198 °C(388 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00012829 CAS Number: 7379-35-3 EINECS: 230-946-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...