1,3-Dibromo-5,5-dimethylhydantoin 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,3-Dibromo-5,5-dimethylimidazolidine-2,4-dione
A15510-0E
A15510-22
A15510-36
CAAAA15510-0EEA
388.68
CAD
CAAAA15510-0E
CAAAA15510-22
CAAAA15510-36
1,3-Dibromo-5,5-dimethylhydantoin 98%
1,3-Dibromo-5,5-dimethylhydantoin
Fieser: 1,208 2,108 12,158 15,112 20,131
Purity: 98%
CAS No.: 77-48-5
Molecular Formula: C5H6Br2N2O2
Formula Weight: 285.93
Flash Point: 155°(311°F)
Melting Point: ca 198° dec.
Beilstein Reference: 146024
UN No.: 3085
MDL No.: MFCD00003189
Formula:
C₅H₆Br₂N₂O₂ Melting Pt: ca 198 °C (dec.) Flash Pt: 155 °C(311 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003189 CAS Number: 77-48-5 EINECS: 201-030-9 UN: 3085 ADR: "5.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...