Diheptyl phthalate 98+%

Supplier: Thermo Scientific Chemicals

A15466-09 A15466-18
CAAAA15466-09EA 110.28 CAD
CAAAA15466-09 CAAAA15466-18 CAAAA15466-30
Diheptyl phthalate 98+%
Diheptyl phthalate

Purity: 98+% CAS No.: 3648-21-3 Molecular Formula: C22H34O4 Formula Weight: 362.51 Flash Point: 213°(415°F) Refractive Index: 1.4860 Boiling Point: 360° Beilstein Reference: 2336623 MDL No.: MFCD00043680

Formula: C₂₂H₃₄O₄
Boiling Pt: 360 °C
Density: 0.992
Flash Pt: 213 °C(415 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043680
CAS Number: 3648-21-3
EINECS: 222-885-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR