3-Chloropropyldimethylammonium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Dimethylaminopropylchloride hydrochloride, 3-Dimethylaminopropyl chloride hydrochloride, N-(3-Chloropropyl)-N,N-dimethylammonium chloride, 3-Chloro-N,N-dimethylpropan-1-amine hydrochloride
A15459-0E
A15459-36
CAAAA15459-0EEA
793.16
CAD
CAAAA15459-0E
CAAAA15459-36
3-Chloropropyldimethylammonium chloride 98%
3-Chloropropyldimethylammonium chloride
Purity: 98%
CAS No.: 5407-04-5
Molecular Formula: Cl(CH2)3N(CH3)2.HCl
Formula Weight: 158.07
Melting Point: 138-144°
Beilstein Reference: 3670046
Formula:
C₅H₁₃Cl₂N MW: 158.07 g/mol Melting Pt: 138…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012521 CAS Number: 5407-04-5 EINECS: 226-467-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...