3-Amino-1H-1,2,4-triazole-5-thiol 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15242-06 A15242-14 A15242-22
CAAAA15242-06EA 53.09 CAD
CAAAA15242-06 CAAAA15242-14 CAAAA15242-22
3-Amino-1H-1,2,4-triazole-5-thiol 98+%
3-Amino-1H-1,2,4-triazole-5-thiol

Purity: 98+% CAS No.: 16691-43-3 Molecular Formula: C2H4N4S Formula Weight: 116.15 Odor: Odorless Melting Point: 300° dec. Beilstein Reference: 112404 MDL No.: MFCD00005231

Formula: C₂H₄N₄S
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005231
CAS Number: 16691-43-3
EINECS: 240-735-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR