Methyl-5-nitro-2-furoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15226-06
A15226-14
CAAAA15226-06EA
115.18
CAD
CAAAA15226-06
CAAAA15226-14
Methyl-5-nitro-2-furoate 97%
Methyl 5-nitro-2-furoate
Purity: 97%
CAS No.: 1874-23-3
Molecular Formula: C6H5NO5
Formula Weight: 171.11
Melting Point: 78-82°
Beilstein Reference: 165132
Formula:
C₆H₅NO₅ MW: 171.11 g/mol Melting Pt: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051956 CAS Number: 1874-23-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...