(4-Bromobenzyl)triphenylphosphonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15220-06
A15220-14
CAAAA15220-06EA
109.02
CAD
CAAAA15220-06
CAAAA15220-14
(4-Bromobenzyl)triphenylphosphonium bromide 98%
(4-Bromobenzyl)triphenylphosphonium bromide
Purity: 98%
CAS No.: 51044-13-4
Molecular Formula: C25H21Br2P
Formula Weight: 512.24
Melting Point: 270-273°
Beilstein Reference: 4173386
Formula:
C₂₅H₂₁Br₂P MW: 512.24 g/mol Melting Pt: 270…273 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051922 CAS Number: 51044-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...