2,6-Dimethoxy-p-benzoquinone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dimethoxy-1,4-Benzoquinone

A14936-03 A14936-06 A14936-14
CAAAA14936-03EA 47.4 CAD
CAAAA14936-03 CAAAA14936-06 CAAAA14936-14
2,6-Dimethoxy-p-benzoquinone 98%
2,6-Dimethoxy-p-benzoquinone

Purity: 98% CAS No.: 530-55-2 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 252-257° Beilstein Reference: 2048182 Merck Reference: 14,3225 MDL No.: MFCD00016368

Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 252…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00016368
CAS Number: 530-55-2
EINECS: 208-484-7
Merck Index: 14,3225

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR