N-(Ethoxycarbonyl)phthalimide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 1,3-dioxoisoindoline 2-carboxylate, N-Carbethoxyphthalimide
A14822-14
A14822-22
CAAAA14822-14EA
64.78
CAD
CAAAA14822-14
CAAAA14822-22
CAAAA14822-36
N-(Ethoxycarbonyl)phthalimide 97%
N-(Ethoxycarbonyl)phthalimide
Fieser: 1,111 5,91 9,212
Purity: 97%
CAS No.: 22509-74-6
Molecular Formula: C11H9NO4
Formula Weight: 219.20
Melting Point: 78-84°
Beilstein Reference: 196340
Formula:
C₁₁H₉NO₄ Melting Pt: 78…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005893 CAS Number: 22509-74-6 EINECS: 245-048-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...