6-Nitrocoumarin 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14792-06
A14792-14
CAAAA14792-06EA
76.16
CAD
CAAAA14792-06
CAAAA14792-14
CAAAA14792-22
6-Nitrocoumarin 98+%
6-Nitrocoumarin
Purity: 98+%
CAS No.: 2725-81-7
Molecular Formula: C9H5NO4
Formula Weight: 191.14
Odor: Odorless
Melting Point: 186-190°
Beilstein Reference: 166079
MDL No.: MFCD00016973
Formula:
C₉H₅NO₄ Melting Pt: 186…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016973 CAS Number: 2725-81-7 EINECS: 220-341-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...