1-Formylpiperazine 90+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Piperazinecarboxaldehyde, 1-Piperazinecarbaldehyde
A14757-14
A14757-22
A14757-36
CAAAA14757-14EA
97.01
CAD
CAAAA14757-14
CAAAA14757-22
CAAAA14757-36
1-Formylpiperazine 90+%
1-Formylpiperazine
Purity: 90+%
CAS No.: 7755-92-2
Molecular Formula: C5H10N2O
Formula Weight: 114.15
Storage Temperature: 2°C to 8°C
Flash Point: 102°(216°F)
Refractive Index: 1.5120
Boiling Point: 264-266°
Beilstein Reference: 110295
MDL No.: MFCD00005963
Formula:
C₅H₁₀N₂O Boiling Pt: 264…266 °C Density: 1.111 Flash Pt: 102 °C(216 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005963 CAS Number: 7755-92-2 EINECS: 231-813-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...