4-(Benzyloxy)-3-methoxybenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
A14716-14
A14716-22
CAAAA14716-14EA
89.27
CAD
CAAAA14716-14
CAAAA14716-22
4-(Benzyloxy)-3-methoxybenzaldehyde 98%
4-(Benzyloxy)-3-methoxybenzaldehyde
Purity: 98%
CAS No.: 2426-87-1
Molecular Formula: C15H14O3
Formula Weight: 242.27
Melting Point: 61-65°
Boiling Point: 213-215°/3.5mm
MDL No.: MFCD00003365
Formula:
C₁₅H₁₄O₃ Boiling Pt: 213…215 °C/3.5 mm Melting Pt: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003365 CAS Number: 2426-87-1 EINECS: 219-379-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...