Acetonyltriphenylphosphonium chloride 99%

Supplier: Thermo Scientific Chemicals
Warning

A14691-14 A14691-22 A14691-36
CAAAA14691-14EA 66.99 CAD
CAAAA14691-14 CAAAA14691-22 CAAAA14691-36
Acetonyltriphenylphosphonium chloride 99%
Acetonyltriphenylphosphonium chloride

Purity: 99% CAS No.: 1235-21-8 Molecular Formula: C21H20ClOP Formula Weight: 354.82 Melting Point: 244-248° MDL No.: MFCD00011813

Formula: C₂₁H₂₀ClOP
Melting Pt: 244…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00011813
CAS Number: 1235-21-8
EINECS: 214-974-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR