Thiophene-2-carbonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14641-14
A14641-22
A14641-36
CAAAA14641-14EA
51.82
CAD
CAAAA14641-14
CAAAA14641-22
CAAAA14641-36
Thiophene-2-carbonyl chloride 98%
Thiophene-2-carbonyl chloride
Purity: 98%
CAS No.: 5271-67-0
Molecular Formula: C5H3ClOS
Formula Weight: 146.60
Storage Temperature: 2°C to 8°C
Flash Point: 90°(194°F)
Refractive Index: 1.5900
Boiling Point: 105°/30mm
Beilstein Reference: 110145
UN No.: 3265
MDL No.: MFCD00005428
Formula:
C₅H₃ClOS Boiling Pt: 105 °C/30 mm Density: 1.372 Flash Pt: 90 °C(194 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005428 CAS Number: 5271-67-0 EINECS: 226-092-4 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...