3-Amino-1H-pyrazole ≥98%

Supplier: Thermo Scientific Chemicals
Danger

A14458-03 A14458-06 A14458-14
CAAAA14458-03EA 59.41 CAD
CAAAA14458-03 CAAAA14458-06 CAAAA14458-14
3-Amino-1H-pyrazole ≥98%
3-Amino-1H-pyrazole

Purity: 97+% CAS No.: 1820-80-0 Molecular Formula: C3H5N3 Formula Weight: 83.09 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 34-39° Boiling Point: 218°/122mm Beilstein Reference: 1618 UN No.: 3259 MDL No.: MFCD00005236

Formula: C₃H₅N₃
Boiling Pt: 218 °C/122 mm
Melting Pt: 34…39 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005236
CAS Number: 1820-80-0
EINECS: 217-338-1
UN: 3263
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR