2-Chloroethylammonium chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14455-0B
A14455-18
A14455-30
CAAAA14455-0BEA
677.82
CAD
CAAAA14455-0B
CAAAA14455-18
CAAAA14455-30
2-Chloroethylammonium chloride 98+%
2-Chloroethylammonium chloride
Purity: 98+%
CAS No.: 870-24-6
Molecular Formula: ClCH2CH2NH2.HCl
Formula Weight: 115.99
Odor: Odorless
pH: 2-3
Melting Point: 143-148°
Beilstein Reference: 3590304
UN No.: 3261
MDL No.: MFCD00012887
Formula:
C₂H₇Cl₂N Melting Pt: 143…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012887 CAS Number: 870-24-6 EINECS: 212-793-2 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...