Oxetane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylene oxide, 1,3-Trimethylene Oxide

A14452-06 A14452-14 A14452-22
CAAAA14452-06EA 259.12 CAD
CAAAA14452-06 CAAAA14452-14 CAAAA14452-22
Oxetane 97%
Oxetane

Fieser: 1,1232


Purity: 97% CAS No.: 503-30-0 Molecular Formula: C3H6O Formula Weight: 58.08 Storage Temperature: 2°C to 8°C Flash Point: -28°(-18°F) Refractive Index: 1.3930 Melting Point: -97° Boiling Point: 49-50° Beilstein Reference: 102382 Merck Reference: 14,9715 UN No.: 1993 MDL No.: MFCD00005167

Formula: C₃H₆O
MW: 58.08
Boiling Pt: 49…50 °C
Melting Pt: –97 °C
Density: 0.891
Flash Pt: –28 °C (–18 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005167
CAS Number: 503-30-0
EINECS: 207-964-3
UN: 1993
ADR: 3 III
Merck Index: 14,9715

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR